3-(4-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione 97%

Supplier: Ambeed
Warning

A1173037-100MG A1173037-1G A1173037-250MG A1173037-25G A1173037-5G
76676-160EA 16.98 USD
76676-160 76676-162 76676-164 76676-166 76676-168
3-(4-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione 97%
3-(4-Bromo-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrN₂O₃
MW: 323.14 g/mol
MDL Number: MFCD32063479
CAS Number: 2093387-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR