N-(trans-4-(3-(2-Chlorophenyl)-7-((3-methyl-4-(4-methylpiperazin-1-yl)phenyl)amino)-2-oxo-3,4-dihydropyrimido[4,5-d]pyrimidin-1(2H)-yl)cyclohexyl)propionamide 99+%
Supplier: Ambeed
|
Warning
|
A1172896-10MG
A1172896-1MG
A1172896-5MG
76676-152EA
184
USD
76676-152
76676-154
76676-156
N-(trans-4-(3-(2-Chlorophenyl)-7-((3-methyl-4-(4-methylpiperazin-1-yl)phenyl)amino)-2-oxo-3,4-dihydropyrimido[4,5-d]pyrimidin-1(2H)-yl)cyclohexyl)propionamide 99+%
N-(trans-4-(3-(2-Chlorophenyl)-7-((3-methyl-4-(4-methylpiperazin-1-yl)phenyl)amino)-2-oxo-3,4-dihydropyrimido[4,5-d]pyrimidin-1(2H)-yl)cyclohexyl)propionamide
Precaución: for Research Use Only
Formula:
C₃₃H₄₁ClN₈O₂ |
MDL Number:
MFCD32644591 CAS Number: 2260886-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...