(R)-3-(Benzhydryloxy)-1-(2-(benzo[d][1,3]dioxol-5-yl)ethyl)piperidine fumarate 98%
Supplier: Ambeed
|
Warning
|
A1169617-10MG
A1169617-1MG
A1169617-50MG
76676-146EA
25
USD
76676-146
76676-148
76676-150
(R)-3-(Benzhydryloxy)-1-(2-(benzo[d][1,3]dioxol-5-yl)ethyl)piperidine fumarate 98%
(R)-3-(Benzhydryloxy)-1-(2-(benzo[d][1,3]dioxol-5-yl)ethyl)piperidine fumarate
Precaución: for Research Use Only
Formula:
C₃₁H₃₃NO₇ |
MDL Number:
MFCD09263617 CAS Number: 127308-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...