1-(Piperidin-4-yl)azetidin-3-ol hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A1169520-100MG
A1169520-1G
A1169520-250MG
76676-140EA
110
USD
76676-140
76676-142
76676-144
1-(Piperidin-4-yl)azetidin-3-ol hydrochloride 98%
1-(Piperidin-4-yl)azetidin-3-ol hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₇ClN₂O |
CAS Number:
1443624-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...