GHK-Cu acetate 98%

Supplier: Ambeed
Warning

Synonyms: N2-(N-glycyl-L-histidyl)-L-Lysine, copper complex,acetate

A1168531-100MG A1168531-1G A1168531-250MG A1168531-5G
76676-108EA 45.62 USD
76676-108 76676-110 76676-112 77620-406
GHK-Cu acetate 98%
GHK-Cu acetate

Precaución: for Research Use Only

Formula: C₁₆H₂₆CuN₆O₆
MW: 461.9602 g/mol
CAS Number: 300801-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR