2,5-Bis(bromomethyl)terephthalonitrile 97%

Supplier: Ambeed
Danger

A1168458-100MG A1168458-250MG
76676-102EA 93.36 USD
76676-102 76676-104
2,5-Bis(bromomethyl)terephthalonitrile 97%
2,5-Bis(bromomethyl)terephthalonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂N₂
MW: 313.9760 g/mol
CAS Number: 64746-04-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR