5-(Ethoxycarbonyl)-3,6-dimethylpyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A1168442-100MG A1168442-250MG
76676-098EA 84 USD
76676-098 76676-100
5-(Ethoxycarbonyl)-3,6-dimethylpyrazine-2-carboxylic acid 97%
5-(Ethoxycarbonyl)-3,6-dimethylpyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₄
CAS Number: 2091155-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR