Zinc(II) pivalate 98%

Supplier: Ambeed
Warning

A1168423-100G A1168423-25G A1168423-5G
76676-092EA 62 USD
76676-092 76676-094 76676-096
Zinc(II) pivalate 98%
Zinc(II) pivalate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄Zn
MW: 267.6565 g/mol
MDL Number: MFCD00082162
CAS Number: 15827-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR