Copper(II) tosylate 98%

Supplier: Ambeed
Danger

A1168407-1G A1168407-5G
76676-078EA 482.71 USD
76676-078 76676-080 76676-082
Copper(II) tosylate 98%
Copper(II) tosylate

Precaución: for Research Use Only

Formula: C₁₄H₁₄CuO₆S₂
MW: 405.9334 g/mol
MDL Number: MFCD16876470
CAS Number: 7144-37-8
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR