18-Bromooctadecanoic acid 98%

Supplier: Ambeed
Warning

A1168394-1G A1168394-5G A1168394-250MG A1168394-25G A1168394-100MG
76676-074EA 95 USD
76676-074 76676-076 77596-000 77596-002 77620-402
18-Bromooctadecanoic acid 98%
18-Bromooctadecanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₅BrO₂
MDL Number: MFCD22055894
CAS Number: 2536-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR