2,7,12-Triamino-5H-diindeno[1,2-a:1',2'-c]fluorene-5,10,15-trione 97%
Supplier: Ambeed
|
Warning
|
A1167927-250MG
A1167927-100MG
76676-056EA
159.14
USD
76676-056
77620-398
2,7,12-Triamino-5H-diindeno[1,2-a:1',2'-c]fluorene-5,10,15-trione 97%
2,7,12-Triamino-5H-diindeno[1,2-a:1',2'-c]fluorene-5,10,15-trione
Precaución: for Research Use Only
Formula:
C₂₇H₁₅N₃O₃ MW: 429.4300 g/mol |
CAS Number:
209112-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...