5-Chloro-3-fluoro-2-iodoaniline 98%

Supplier: Ambeed
Warning

A1167750-100MG A1167750-1G A1167750-250MG A1167750-5G
76676-046EA 32.89 USD
76676-046 76676-048 76676-050 76676-052
5-Chloro-3-fluoro-2-iodoaniline 98%
5-Chloro-3-fluoro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄ClFIN
MW: 271.4585 g/mol
CAS Number: 2092184-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR