tert-Butyl (S)-2-(cyanomethyl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A1167527-100MG A1167527-1G A1167527-250MG A1167527-5G
76676-034EA 31.83 USD
76676-034 76676-036 76676-038 77549-330
tert-Butyl (S)-2-(cyanomethyl)piperazine-1-carboxylate 98%
tert-Butyl (S)-2-(cyanomethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉N₃O₂
MW: 225.2875 g/mol
MDL Number: MFCD32263269
CAS Number: 1589565-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR