(S)-2-((2-Ethoxyphenoxy)methyl)morpholine hydrochloride 98%

Supplier: Ambeed
Warning

A1167355-10MG A1167355-1MG A1167355-25MG A1167355-5MG
76676-026EA 51 USD
76676-026 76676-028 76676-030 76676-032
(S)-2-((2-Ethoxyphenoxy)methyl)morpholine hydrochloride 98%
(S)-2-((2-Ethoxyphenoxy)methyl)morpholine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₀ClNO₃
CAS Number: 56287-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR