(R)-2-((2-Ethoxyphenoxy)methyl)morpholine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Viloxazine HCl

A1167354-10MG A1167354-1MG A1167354-25MG A1167354-50MG A1167354-5MG
76676-016EA 82.76 USD
76676-016 76676-018 76676-020 76676-022 76676-024
(R)-2-((2-Ethoxyphenoxy)methyl)morpholine hydrochloride 98%
(R)-2-((2-Ethoxyphenoxy)methyl)morpholine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₀ClNO₃
MW: 273.7558 g/mol
MDL Number: MFCD18379655
CAS Number: 56287-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR