(R)-4-(tert-Butyl)-N-(6-(2,3-dihydroxypropoxy)-5-(2-methoxyphenoxy)-2-(4-methoxyphenyl)pyrimidin-4-yl)benzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: Ro 46-8443

A1167274-10MG A1167274-1MG A1167274-25MG A1167274-50MG A1167274-5MG
76676-006EA 49.87 USD
76676-006 76676-008 76676-010 76676-012 76676-014
(R)-4-(tert-Butyl)-N-(6-(2,3-dihydroxypropoxy)-5-(2-methoxyphenoxy)-2-(4-methoxyphenyl)pyrimidin-4-yl)benzenesulfonamide 98%
(R)-4-(tert-Butyl)-N-(6-(2,3-dihydroxypropoxy)-5-(2-methoxyphenoxy)-2-(4-methoxyphenyl)pyrimidin-4-yl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₃₁H₃₅N₃O₈S
MW: 609.69 g/mol
MDL Number: MFCD00946586
CAS Number: 175556-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR