(R)-6,7-Dimethoxy-2-methyl-N-(1-(4-(2-((methylamino)methyl)phenyl)thiophen-2-yl)ethyl)quinazolin-4-amine 98%

Supplier: Ambeed
Warning

Synonyms: BAY-293

A1167268-10MG A1167268-1MG A1167268-50MG
76676-000EA 84 USD
76676-000 76676-002 76676-004
(R)-6,7-Dimethoxy-2-methyl-N-(1-(4-(2-((methylamino)methyl)phenyl)thiophen-2-yl)ethyl)quinazolin-4-amine 98%
(R)-6,7-Dimethoxy-2-methyl-N-(1-(4-(2-((methylamino)methyl)phenyl)thiophen-2-yl)ethyl)quinazolin-4-amine

Precaución: for Research Use Only

Formula: C₂₅H₂₈N₄O₂S
MW: 448.58 g/mol
MDL Number: MFCD31813755
CAS Number: 2244904-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR