(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol 2-(3,5-dichlorophenyl)benzo[d]oxazole-6-carboxylate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Tafamidis meglumine
A1167264-10MG
A1167264-1MG
A1167264-25MG
A1167264-5MG
76675-986EA
78.51
USD
76675-986
76675-988
76675-990
76675-992
(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol 2-(3,5-dichlorophenyl)benzo[d]oxazole-6-carboxylate 98%
(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol 2-(3,5-dichlorophenyl)benzo[d]oxazole-6-carboxylate
Precaución: for Research Use Only
Formula:
C₂₁H₂₄Cl₂N₂O₈ MW: 503.33 g/mol |
MDL Number:
MFCD28502032 CAS Number: 951395-08-7 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...