N-(2-Aminoethyl)-2-(4-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)phenoxy)acetamide hydrochloride 98% (contains ~trihydrochloride)

Supplier: Ambeed
Warning

Synonyms: Xanthine amine congener HCl

A1167146-10MG A1167146-1MG A1167146-25MG A1167146-5MG
76675-978EA 84 USD
76675-978 76675-980 76675-982 76675-984
N-(2-Aminoethyl)-2-(4-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)phenoxy)acetamide hydrochloride 98% (contains ~trihydrochloride)
N-(2-Aminoethyl)-2-(4-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)phenoxy)acetamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₉ClN₆O₄
MDL Number: MFCD00055115
CAS Number: 1783977-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR