1-([1,1'-Biphenyl]-3-yl)-4-amino-5-chloropyrimidin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: Bobcat339

A1167136-10MG A1167136-25MG A1167136-50MG A1167136-5MG A1167136-1MG
76675-970EA 83.82 USD
76675-970 76675-972 76675-974 76675-976 77620-378
1-([1,1'-Biphenyl]-3-yl)-4-amino-5-chloropyrimidin-2(1H)-one 98%
1-([1,1'-Biphenyl]-3-yl)-4-amino-5-chloropyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₆H₁₂ClN₃O
MW: 297.74 g/mol
MDL Number: MFCD32644557
CAS Number: 2280037-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR