6,6′-Bis(4-(S)-isopropyl-2-oxazolinyl)-2,2′-bipyridine 98%

Supplier: Ambeed
Warning

A1167084-100MG A1167084-1G A1167084-250MG
76675-960EA 77 USD
76675-960 76675-962 76675-964
6,6′-Bis(4-(S)-isopropyl-2-oxazolinyl)-2,2′-bipyridine 98%
6,6′-Bis(4-(S)-isopropyl-2-oxazolinyl)-2,2′-bipyridine

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₄O₂
MDL Number: MFCD22417223
CAS Number: 147409-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR