Iron tris(diisobutyrylmethane) 98%

Supplier: Ambeed
Warning

A1167075-100MG A1167075-1G A1167075-250MG A1167075-5G
76675-952EA 23 USD
76675-952 76675-954 76675-956 76675-958
Iron tris(diisobutyrylmethane) 98%
Iron tris(diisobutyrylmethane)

Precaución: for Research Use Only

MDL Number: MFCD28346980
CAS Number: 24444-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR