[4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine] nickel (II) dichloride 97%

Supplier: Ambeed
Danger

A1167069-100MG A1167069-1G A1167069-5G A1167069-250MG A1167069-25G
76675-950EA 10 USD
76675-950 77297-492 77549-326 77566-120 77866-211
[4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine] nickel (II) dichloride 97%
[4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine] nickel (II) dichloride

Precaución: For research use only.

Formula: C₁₈H₂₄Cl₂N₂Ni
MW: 398.0000 g/mol
MDL Number: MFCD31807325
CAS Number: 1034901-50-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR