(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((tert-butoxycarbonyl)(methyl)amino)propanoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Dap(Boc,Me)-OH
A1167050-1G
A1167050-5G
76675-944EA
1167
USD
76675-944
77866-209
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((tert-butoxycarbonyl)(methyl)amino)propanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((tert-butoxycarbonyl)(methyl)amino)propanoic acid
Precaución: for Research Use Only
Formula:
C₂₄H₂₈N₂O₆ MW: 440.49 g/mol |
MDL Number:
MFCD31380696 CAS Number: 446847-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...