N-((R)-sec-Butyl)-8-(6-methoxy-2-methylpyridin-3-yl)-2,7-dimethylpyrazolo[1,5-a][1,3,5]triazin-4-amine 98%

Supplier: Ambeed
Warning

Synonyms: Pexacerfont

A1166938-10MG A1166938-5MG
76675-940EA 309.79 USD
76675-940 76675-942
N-((R)-sec-Butyl)-8-(6-methoxy-2-methylpyridin-3-yl)-2,7-dimethylpyrazolo[1,5-a][1,3,5]triazin-4-amine 98%
N-((R)-sec-Butyl)-8-(6-methoxy-2-methylpyridin-3-yl)-2,7-dimethylpyrazolo[1,5-a][1,3,5]triazin-4-amine

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₆O
MW: 340.42 g/mol
MDL Number: MFCD22628864
CAS Number: 459856-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR