Nickelbis(trifluoromethylsulfonyl)imide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Nickel bis(trifluoromethanesulphonimide), Nickel bis(trifluoromethanesulphonyl)imide, Nickel bis(trifluoromethylsulphonyl)imide
A1166525-1G
A1166525-250MG
76675-922EA
103.97
USD
76675-922
76675-924
Nickelbis(trifluoromethylsulfonyl)imide 98%
Nickelbis(trifluoromethylsulfonyl)imide
Precaución: for Research Use Only
Formula:
C₄F₁₂N₂NiO₈S₄ MW: 618.96 g/mol |
MDL Number:
MFCD23380174 CAS Number: 207861-63-0 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...