Cobalt bis(trifluoromethylsulfonyl)imide 98%

Supplier: Ambeed
Danger

Synonyms: Bis(trifluoromethanesulphonyl)imide cobalt salt, Bis(trifluoromethane)sulphonimide cobalt salt

A1166524-1G A1166524-5G A1166524-250MG A1166524-25G
76675-918EA 61 USD
76675-918 76675-920 77595-986 77866-206
Cobalt bis(trifluoromethylsulfonyl)imide 98%
Cobalt bis(trifluoromethylsulfonyl)imide

Precaución: for Research Use Only

Formula: C₄CoF₁₂N₂O₈S₄
MW: 619.23 g/mol
MDL Number: MFCD23380169
CAS Number: 207861-61-8
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR