5-Hydroxypentyl acrylate 92% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with hydroquinone (HQ)

A1166445-1G A1166445-250MG A1166445-5G
76675-908EA 82.76 USD
76675-908 76675-910 76675-912
5-Hydroxypentyl acrylate 92% stabilized
5-Hydroxypentyl acrylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.1950 g/mol
MDL Number: MFCD18974086
CAS Number: 57198-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR