(2-Hydroxybenzyl)diphenylphosphine oxide 98%

Supplier: Ambeed
Warning

A1166439-100G A1166439-10G A1166439-1G A1166439-250MG A1166439-5G A1166439-25G
76675-898EA 1141.53 USD
76675-898 76675-900 76675-902 76675-904 76675-906 77549-322
(2-Hydroxybenzyl)diphenylphosphine oxide 98%
(2-Hydroxybenzyl)diphenylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₉H₁₇O₂P
MW: 308.3109 g/mol
MDL Number: MFCD00406515
CAS Number: 70127-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR