1-(3-(1H-Imidazol-1-yl)propyl)-3-(3,4-dimethoxyphenyl)thiourea 98%

Supplier: Ambeed
Warning

Synonyms: PBD-150

A1166381-10MG A1166381-25MG A1166381-5MG A1166381-1MG
76675-888EA 58.35 USD
76675-888 76675-890 76675-892 77595-984
1-(3-(1H-Imidazol-1-yl)propyl)-3-(3,4-dimethoxyphenyl)thiourea 98%
1-(3-(1H-Imidazol-1-yl)propyl)-3-(3,4-dimethoxyphenyl)thiourea

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₄O₂S
MW: 320.41 g/mol
MDL Number: MFCD32666996
CAS Number: 790663-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR