(S)-2-Amino-4-((tert-butyldimethylsilyl)oxy)butanoic acid 97%

Supplier: Ambeed
Warning

A1166155-100MG A1166155-1G A1166155-250MG A1166155-5G
76675-880EA 10.61 USD
76675-880 76675-882 76675-884 76675-886
(S)-2-Amino-4-((tert-butyldimethylsilyl)oxy)butanoic acid 97%
(S)-2-Amino-4-((tert-butyldimethylsilyl)oxy)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₂₃NO₃Si
MW: 233.3800 g/mol
MDL Number: MFCD23164182
CAS Number: 474023-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR