5-(4-Bromophenyl)-1,3,4-thiadiazole-2(3H)-thione 97%

Supplier: Ambeed
Warning

A1166127-100MG A1166127-250MG
76675-876EA 176.11 USD
76675-876 76675-878
5-(4-Bromophenyl)-1,3,4-thiadiazole-2(3H)-thione 97%
5-(4-Bromophenyl)-1,3,4-thiadiazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₈H₅BrN₂S₂
MW: 273.1727 g/mol
MDL Number: MFCD08702338
CAS Number: 1021953-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR