Tetrabutylammonium trifluoromethanesulfonate 97%
Supplier: Ambeed
|
Warning
|
A1165828-100G
A1165828-25G
A1165828-5G
A1165828-500G
76675-830EA
104
USD
76675-830
76675-832
76675-834
77866-201
Tetrabutylammonium trifluoromethanesulfonate 97%
Tetrabutylammonium trifluoromethanesulphonate
Precaución: for Research Use Only
Formula:
C₁₇H₃₆F₃NO₃S |
MDL Number:
MFCD00042585 CAS Number: 35895-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...