Iridium(1+), [4,4′-bis(1,1-dimethylethyl)-2,2′-bipyridine-κN1,κN1′]bis[5-(1,1-dimethylethyl)-2-(2-pyridinyl-κN)phenyl-κC]-, hexafluorophosphate(1-) (1:1) 98%

Supplier: Ambeed
Warning

A1165826-100MG A1165826-1G A1165826-50MG A1165826-250MG
76675-824EA 68 USD
76675-824 76675-826 76675-828 77866-200
Iridium(1+), [4,4′-bis(1,1-dimethylethyl)-2,2′-bipyridine-κN1,κN1′]bis[5-(1,1-dimethylethyl)-2-(2-pyridinyl-κN)phenyl-κC]-, hexafluorophosphate(1-) (1:1) 98%
Iridium(1+), [4,4′-bis(1,1-dimethylethyl)-2,2′-bipyridine-κN1,κN1′]bis[5-(1,1-dimethylethyl)-2-(2-pyridinyl-κN)phenyl-κC]-, hexafluorophosphate(1-) (1:1)

Precaución: for Research Use Only

Formula: C₄₈H₅₆F₆IrN₄P
MW: 1026.1662 g/mol
CAS Number: 1570275-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR