N-(2-(3-(Piperazin-1-ylmethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)quinoxaline-2-carboxamide xhydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: SRT1720, SRT 1720 hydrochloride

A1165814-1MG A1165814-25MG A1165814-1G A1165814-100MG A1165814-250MG A1165814-50MG
76675-820EA 10.61 USD
76675-820 76675-822 77566-110 77595-982 77620-356 77620-358
N-(2-(3-(Piperazin-1-ylmethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)quinoxaline-2-carboxamide xhydrochloride 98%
N-(2-(3-(Piperazin-1-ylmethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)quinoxaline-2-carboxamide xhydrochloride

Precaución: For research use only.

Formula: C₂₅H₂₃N₇OS·HCl
MW: 506.02 g/mol
MDL Number: MFCD18074509
CAS Number: 1001645-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR