2-Hydroxy-2-(4-(trifluoromethoxy)phenyl)acetonitrile 97%

Supplier: Ambeed
Warning

A1165583-100MG A1165583-1G A1165583-250MG
76675-808EA 33.95 USD
76675-808 76675-810 76675-812
2-Hydroxy-2-(4-(trifluoromethoxy)phenyl)acetonitrile 97%
2-Hydroxy-2-(4-(trifluoromethoxy)phenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₆F₃NO₂
MW: 217.1447 g/mol
MDL Number: MFCD12169348
CAS Number: 1155915-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR