N,N'-Di-n-octyl-3,4,9,10-perylenetetracarboxylic diimide 97%

Supplier: Ambeed
Warning

Synonyms: PTCDI-C8, 2,9-Dioctylanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone

A1165551-100MG A1165551-1G
76675-802EA 27.59 USD
76675-802 76675-804
N,N'-Di-n-octyl-3,4,9,10-perylenetetracarboxylic diimide 97%
N,N'-Di-n-octyl-3,4,9,10-perylenetetracarboxylic diimide

Precaución: for Research Use Only

Formula: C₄₀H₄₂N₂O₄
MW: 614.79 g/mol
MDL Number: MFCD08276854
CAS Number: 78151-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR