N1,N3-DiBoc-spermidine 97%

Supplier: Ambeed
Warning

A1165501-100MG A1165501-250MG
76675-798EA 52 USD
76675-798 76675-800
N1,N3-DiBoc-spermidine 97%
N1,N3-DiBoc-spermidine

Precaución: for Research Use Only

Formula: C₁₇H₃₅N₃O₄
MW: 345.4775 g/mol
CAS Number: 83392-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR