3,6-Di-tert-butylacridin-9(10H)-one 97%

Supplier: Ambeed
Warning

A1165300-100MG A1165300-1G A1165300-250MG
76675-784EA 83 USD
76675-784 76675-786 76675-788
3,6-Di-tert-butylacridin-9(10H)-one 97%
3,6-Di-tert-butylacridin-9(10H)-one

Precaución: for Research Use Only

Formula: C₂₁H₂₅NO
CAS Number: 1810004-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR