1,8-Dibromo-3,6-di-tert-butyl-9H-carbazole 97%

Supplier: Ambeed
Warning

A1165256-1G A1165256-25G A1165256-5G
76675-774EA 13 USD
76675-774 76675-776 77549-316
1,8-Dibromo-3,6-di-tert-butyl-9H-carbazole 97%
1,8-Dibromo-3,6-di-tert-butyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₀H₂₃Br₂N
MDL Number: MFCD31692862
CAS Number: 625385-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR