(1S,2R,3R,4S)-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A1165252-100G A1165252-25G
76675-770EA 54 USD
76675-770 76675-772 77571-004
(1S,2R,3R,4S)-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid 97%
(1S,2R,3R,4S)-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MDL Number: MFCD12026879
CAS Number: 3853-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR