(Z)-4-(Octadecyloxy)-4-oxobut-2-enoic acid 98%

Supplier: Ambeed
Warning

A1165228-10G A1165228-1G A1165228-5G
76675-764EA 345.86 USD
76675-764 76675-766 76675-768
(Z)-4-(Octadecyloxy)-4-oxobut-2-enoic acid 98%
(Z)-4-(Octadecyloxy)-4-oxobut-2-enoic acid

Precaución: for Research Use Only

Formula: C₂₂H₄₀O₄
MW: 368.5506 g/mol
MDL Number: MFCD08274815
CAS Number: 2424-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR