Phen-NaDPO 98%

Supplier: Ambeed
Warning

A1165130-100MG A1165130-250MG
76675-760EA 220.67 USD
76675-760 76675-762
Phen-NaDPO 98%
Phen-NaDPO

Precaución: for Research Use Only

Formula: C₃₄H₂₃N₂OP
MW: 506.5330 g/mol
MDL Number: MFCD31811570
CAS Number: 1480371-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR