1-(2-Ethylpyrimidin-5-yl)ethanone 97%

Supplier: Ambeed
Warning

A1165110-100MG A1165110-250MG
76675-756EA 47.75 USD
76675-756 76675-758
1-(2-Ethylpyrimidin-5-yl)ethanone 97%
1-(2-Ethylpyrimidin-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
MDL Number: MFCD26521274
CAS Number: 1511328-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR