(4S,4'S)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) 97%

Supplier: Ambeed
Warning

A1165106-1G A1165106-250MG
76675-752EA 528.33 USD
76675-752 76675-754
(4S,4'S)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) 97%
(4S,4'S)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₃₅H₃₄N₂O₂
MW: 514.6567 g/mol
CAS Number: 583058-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR