(3aR,3a'R,8aS,8a'S)-2,2'-(Cyclopropane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole) ≥98%, ee 99%

Supplier: Ambeed
Warning

A1165102-100MG A1165102-1G A1165102-250MG
76675-746EA 20 USD
76675-746 76675-748 76675-750
(3aR,3a'R,8aS,8a'S)-2,2'-(Cyclopropane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole) ≥98%, ee 99%
(3aR,3a'R,8aS,8a'S)-2,2'-(Cyclopropane-1,1-diyl)bis(8,8a-dihydro-3aH-indeno[1,2-d]oxazole)

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₂O₂
MDL Number: MFCD32641142
CAS Number: 229184-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR