N-((6-Fluoro-2,3,4,9-tetrahydro-1H-carbazol-3-yl)methyl)-4,4-bis(4-fluorophenyl)butan-1-amine 98%

Supplier: Ambeed
Warning

Synonyms: MSC1094308

A1164830-1MG A1164830-5MG
76675-722EA 83.82 USD
76675-722 76675-724
N-((6-Fluoro-2,3,4,9-tetrahydro-1H-carbazol-3-yl)methyl)-4,4-bis(4-fluorophenyl)butan-1-amine 98%
N-((6-Fluoro-2,3,4,9-tetrahydro-1H-carbazol-3-yl)methyl)-4,4-bis(4-fluorophenyl)butan-1-amine

Precaución: for Research Use Only

Formula: C₂₉H₂₉F₃N₂
MW: 462.5492 g/mol
MDL Number: MFCD31814427
CAS Number: 2219320-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR