4-(2-Bromo-6-fluorobenzoyl)-N-(pyridin-3-yl)-1H-pyrrole-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: ERK5-IN-2

A1164824-10MG A1164824-1MG A1164824-25MG A1164824-50MG A1164824-5MG
76675-712EA 35.01 USD
76675-712 76675-714 76675-716 76675-718 76675-720
4-(2-Bromo-6-fluorobenzoyl)-N-(pyridin-3-yl)-1H-pyrrole-2-carboxamide 98%
4-(2-Bromo-6-fluorobenzoyl)-N-(pyridin-3-yl)-1H-pyrrole-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₁₁BrFN₃O₂
MW: 388.19 g/mol
MDL Number: MFCD32174287
CAS Number: 1888305-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR