1,2-Ethanedisulfonic acid ≥98% (contains < 20% H₂O)

Supplier: Ambeed
Danger

Synonyms: Ethane-1,2-disulphonic acid

A1164802-5G A1164802-25G
76675-702EA 16.98 USD
76675-702 77571-000
1,2-Ethanedisulfonic acid ≥98% (contains < 20% H₂O)
1,2-Ethanedisulphonic acid

Precaución: for Research Use Only

Formula: C₂H₆O₆S₂
MW: 190.19 g/mol
MDL Number: MFCD00069901
CAS Number: 110-04-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR