(E)-tert-Butyldimethyl((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)allyl)oxy)silane 95%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(tert-Butyldimethylsilyloxy)propene-1-yl-boronic acid pinacol ester

A1164792-100MG A1164792-1G A1164792-250MG A1164792-5G
76675-694EA 46.68 USD
76675-694 76675-696 76675-698 76675-700
(E)-tert-Butyldimethyl((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)allyl)oxy)silane 95%
(E)-tert-Butyldimethyl((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)allyl)oxy)silane

Precaución: for Research Use Only

Formula: C₁₅H₃₁BO₃Si
MW: 298.3 g/mol
MDL Number: MFCD15144803
CAS Number: 114653-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR